5-(1-Methylethyl)-2-pyridinamine

APOSOR323123-250MGEA 0 EUR
APOSOR323123-250MG
5-(1-Methylethyl)-2-pyridinamine
5-(1-Methylethyl)-2-pyridinamine
Formel: C₈H₁₂N₂
Molecular Weight: 136,194 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08460246
CAS-Nummer: 603310-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR