5-Ethylpyrimidin-4-ol

Lieferant: Apollo Scientific

APOSOR323119-500MGEA 598 EUR
APOSOR323119-500MG
5-Ethylpyrimidin-4-ol
5-Ethylpyrimidin-4-ol
Formel: C₆H₈N₂O
Molecular Weight: 124,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09750155
CAS-Nummer: 25198-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR