2-(m-Tolylthio)-6-chloropyridine

APOSOR323115-500MGEA 0 EUR
APOSOR323115-500MG
2-(m-Tolylthio)-6-chloropyridine
2-(m-Tolylthio)-6-chloropyridine
Formel: C₁₂H₁₀ClNS
Molecular Weight: 235,733 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16987548
CAS-Nummer: 1242241-04-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR