3-(4-Methylphenoxy)butan-2-one, tech
Lieferant: Apollo Scientific
APOSOR32310-1GEA
1390
EUR
APOSOR32310-1G
3-(4-Methylphenoxy)butan-2-one, tech
3-(4-Methylphenoxy)butan-2-one
Formel:
C₁₁H₁₄O₂ Molecular Weight: 178,228 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00243577 CAS-Nummer: 91969-89-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...