1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-2-propyl-1H-benzo[d]imidazole-5-carboxylic acid

APOSOR323099-500MGEA 0 EUR
APOSOR323099-500MG
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-2-propyl-1H-benzo[d]imidazole-5-carboxylic acid
1-(2,3-Dihydrobenzo[b][1,4]dioxin-6-yl)-2-propyl-1H-benzo[d]imidazole-5-carboxylic acid
Formel: C₁₉H₁₈N₂O₄
Molecular Weight: 338,357 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18157698
CAS-Nummer: 942289-81-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR