2-(2-tert-Butoxycarbonylamino-3-phenyl-propionylamino)-3-phenyl-propionic acid methyl ester

Lieferant: Apollo Scientific

APOSOR323093-250MGEA 587 EUR
APOSOR323093-250MG
2-(2-tert-Butoxycarbonylamino-3-phenyl-propionylamino)-3-phenyl-propionic acid methyl ester
2-(2-tert-Butoxycarbonylamino-3-phenyl-propionylamino)-3-phenyl-propionic acid methyl ester
Formel: C₂₄H₃₀N₂O₅
Molecular Weight: 426,505 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 13122-89-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR