4-(4-Bromo-2-chloro-5-methoxyphenyl)-6-methyl-2-(methylsulphonyl)pyrimidine

APOSOR323091-250MGEA 0 EUR
APOSOR323091-250MG
4-(4-Bromo-2-chloro-5-methoxyphenyl)-6-methyl-2-(methylsulphonyl)pyrimidine
4-(4-Bromo-2-chloro-5-methoxyphenyl)-6-methyl-2-(methylsulphonyl)pyrimidine
Formel: C₁₃H₁₂BrClN₂O₃S
Molecular Weight: 391,668 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18157705
CAS-Nummer: 1258652-61-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR