1-(tert-Butoxycarbonyl)-4-(4-phenyl-1H-1,2,3-triazol-1-yl)pyrrolidine-2-carboxylic acid

APOSOR323088-250MGEA 0 EUR
APOSOR323088-250MG
1-(tert-Butoxycarbonyl)-4-(4-phenyl-1H-1,2,3-triazol-1-yl)pyrrolidine-2-carboxylic acid
1-(tert-Butoxycarbonyl)-4-(4-phenyl-1H-1,2,3-triazol-1-yl)pyrrolidine-2-carboxylic acid
Formel: C₁₈H₂₂N₄O₄
Molecular Weight: 358,392 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18157720
CAS-Nummer: 1258652-60-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR