tert-Butyl (1r,5s,6s)-3-benzyl-3-aza-bicyclo[3.1.0]hexan-6-ylcarbamate

APOSOR323087-250MGEA 0 EUR
APOSOR323087-250MG
tert-Butyl (1r,5s,6s)-3-benzyl-3-aza-bicyclo[3.1.0]hexan-6-ylcarbamate
tert-Butyl (1r,5s,6s)-3-benzyl-3-aza-bicyclo[3.1.0]hexan-6-ylcarbamate
Formel: C₁₇H₂₄N₂O₂
Molecular Weight: 288,385 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16987527
CAS-Nummer: 185559-52-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR