3-(2-Methoxybenzylsulphonyl)pyrrolidine

APOSOR323033-250MGEA 0 EUR
APOSOR323033-250MG
3-(2-Methoxybenzylsulphonyl)pyrrolidine
3-(2-Methoxybenzylsulphonyl)pyrrolidine
Formel: C₁₂H₁₇NO₃S
Molecular Weight: 255,333 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585300
CAS-Nummer: 1206970-68-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR