5-(4-Methoxyphenyl)imidazo[1,5-a]pyrazine-3-carboxylic acid

APOSOR322921-250MGEA 0 EUR
APOSOR322921-250MG
5-(4-Methoxyphenyl)imidazo[1,5-a]pyrazine-3-carboxylic acid
5-(4-Methoxyphenyl)imidazo[1,5-a]pyrazine-3-carboxylic acid
Formel: C₁₄H₁₁N₃O₃
Molecular Weight: 269,255 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585307
CAS-Nummer: 1206969-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR