3-[(4-Chlorophenyl)amino]propanoic acid 95%

Lieferant: Apollo Scientific

APOSOR32288-1GEA 1390 EUR
APOSOR32288-1G
3-[(4-Chlorophenyl)amino]propanoic acid 95%
3-[(4-Chlorophenyl)amino]propanoic acid
Formel: C₉H₁₀ClNO₂
Molecular Weight: 199,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00831705
CAS-Nummer: 21617-19-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR