Biphenyl-4-ylmethyl-piperidin-3-yl-carbamic acid tert-butyl ester

Lieferant: Apollo Scientific

APOSOR322868-250MGEA 441 EUR
APOSOR322868-250MG
Biphenyl-4-ylmethyl-piperidin-3-yl-carbamic acid tert-butyl ester
Biphenyl-4-ylmethyl-piperidin-3-yl-carbamic acid tert-butyl ester
Formel: C₂₃H₃₀N₂O₂
Molecular Weight: 366,497 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585270
CAS-Nummer: 1206969-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR