3,6-Dioxoctanedioic acid bis-methyl ester

APOSOR322795-500MGEA 0 EUR
APOSOR322795-500MG APOSOR322795-1G
3,6-Dioxoctanedioic acid bis-methyl ester
3,6-Dioxoctanedioic acid bis-methyl ester
Formel: C₈H₁₄O₆
Molecular Weight: 206,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01095847
CAS-Nummer: 54665-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR