4-Amino-5-(2-ethoxyphenyl)-4H-1,2,4-triazole-3-thiol

Lieferant: Apollo Scientific

APOSOR322785-500MGEA 367 EUR
APOSOR322785-500MG
4-Amino-5-(2-ethoxyphenyl)-4H-1,2,4-triazole-3-thiol
4-Amino-5-(2-ethoxyphenyl)-4H-1,2,4-triazole-3-thiol
Formel: C₁₀H₁₂N₄OS
Molecular Weight: 236,293 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06655276
CAS-Nummer: 851879-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR