tert-Butyl 2-iodo-1H-imidazole-1-carboxylate
Lieferant: Apollo Scientific
APOSOR322783-250MGEA
367
EUR
APOSOR322783-250MG
tert-Butyl 2-iodo-1H-imidazole-1-carboxylate
tert-Butyl 2-iodo-1H-imidazole-1-carboxylate
Formel:
C₈H₁₁IN₂O₂ Molecular Weight: 294,09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14585432 CAS-Nummer: 181220-75-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...