2-(4-Benzylpiperazin-1-yl)acetohydrazide

Lieferant: Apollo Scientific

APOSOR322782-250MGEA 367 EUR
APOSOR322782-250MG
2-(4-Benzylpiperazin-1-yl)acetohydrazide
2-(4-Benzylpiperazin-1-yl)acetohydrazide
Formel: C₁₃H₂₀N₄O
Molecular Weight: 248,324 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07364291
CAS-Nummer: 24632-70-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR