3,4-Diamino-N-(4-(2-(4-methylpiperazin-1-yl)ethyl)phenyl)benzamide

Lieferant: Apollo Scientific

APOSOR322673-100MGEA 294 EUR
APOSOR322673-100MG
3,4-Diamino-N-(4-(2-(4-methylpiperazin-1-yl)ethyl)phenyl)benzamide
3,4-Diamino-N-(4-(2-(4-methylpiperazin-1-yl)ethyl)phenyl)benzamide
Formel: C₂₀H₂₇N₅O
Molecular Weight: 353,461 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14585347
CAS-Nummer: 936940-80-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR