1-Bromo-3-(Bromomethyl)adamantane

Lieferant: Apollo Scientific

APOSOR322641-100MGEA 274 EUR
APOSOR322641-100MG
1-Bromo-3-(Bromomethyl)adamantane
1-Bromo-3-(Bromomethyl)adamantane
Formel: C₁₁H₁₆Br₂
Molecular Weight: 308,053 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00185299
CAS-Nummer: 1822-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR