(E)-3-(2-Carboxyethylcarbamoyl)acrylic acid

Lieferant: Apollo Scientific

APOSOR322566-1GEA 207 EUR
APOSOR322566-1G
(E)-3-(2-Carboxyethylcarbamoyl)acrylic acid
(E)-3-(2-Carboxyethylcarbamoyl)acrylic acid
Formel: C₇H₉NO₅
Molecular Weight: 187,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00461553
CAS-Nummer: 904217-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR