N-(3-Hydroxyphenyl)formamide

Lieferant: Apollo Scientific

APOSOR322539-100MGEA 200 EUR
APOSOR322539-100MG
N-(3-Hydroxyphenyl)formamide
N-(3-Hydroxyphenyl)formamide
Formel: C₇H₇NO₂
Molecular Weight: 137,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03147263
CAS-Nummer: 24891-35-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR