4-Hydroxypyrrolidin-2-carboxamide

Lieferant: Apollo Scientific

APOSOR322526-100MGEA 190 EUR
APOSOR322526-100MG
4-Hydroxypyrrolidin-2-carboxamide
4-Hydroxypyrrolidin-2-carboxamide
Formel: C₅H₁₀N₂O₂
Molecular Weight: 130,145 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04635892
CAS-Nummer: 1000616-27-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR