5-[(Dimethylamino)methyl]-2-furoic acid

Lieferant: Apollo Scientific

APOSOR322456-100MGEA 263 EUR
APOSOR322456-100MG
5-[(Dimethylamino)methyl]-2-furoic acid
5-[(Dimethylamino)methyl]-2-furoic acid
Formel: C₈H₁₁NO₃
Molecular Weight: 169,178 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03274585
CAS-Nummer: 86649-59-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR