1-(5-Bromo-2-thienyl)methanamine

Achtung

APOSOR322390-250MGEA 0 EUR
APOSOR322390-250MG
1-(5-Bromo-2-thienyl)methanamine
1-(5-Bromo-2-thienyl)methanamine
Formel: C₅H₆BrNS
Molecular Weight: 192,077 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06213820
CAS-Nummer: 612499-04-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR