2-Phenylimidazo[1,2-a]pyrimidine-3-carbaldehyde

Lieferant: Apollo Scientific

APOSOR322289-1GEA 149 EUR
APOSOR322289-1G
2-Phenylimidazo[1,2-a]pyrimidine-3-carbaldehyde
2-Phenylimidazo[1,2-a]pyrimidine-3-carbaldehyde
Formel: C₁₃H₉N₃O
Molecular Weight: 223,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06255242
CAS-Nummer: 74944-29-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR