6-Nitro-2-benzothiazolinone 98%

Lieferant: Apollo Scientific

Achtung

APOSOR322239-5GEA 371 EUR
APOSOR322239-5G APOSOR322239-1G
6-Nitro-2-benzothiazolinone 98%
6-Nitro-2-benzothiazolinone
Formel: C₇H₄N₂O₃S
Molecular Weight: 196,183 g/mol
Schmelzpunkt: 246 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00239350
CAS-Nummer: 28620-12-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR