(2E)-3-(Dimethylamino)-1-(furan-3-yl)prop-2-en-1-one 95%
Lieferant: Apollo Scientific
APOSOR32220-1GEA
1390
EUR
APOSOR32220-1G
(2E)-3-(Dimethylamino)-1-(furan-3-yl)prop-2-en-1-one 95%
(2E)-3-(Dimethylamino)-1-(furan-3-yl)prop-2-en-1-one
Formel:
C₉H₁₁NO₂ Molecular Weight: 165,189 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02082364 CAS-Nummer: 67382-49-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...