4,6-Dichloro-3-formylcoumarin

Lieferant: Apollo Scientific

APOSOR322193-250MGEA 141 EUR
APOSOR322193-250MG
4,6-Dichloro-3-formylcoumarin
4,6-Dichloro-3-formylcoumarin
Formel: C₁₀H₄Cl₂O₃
Molecular Weight: 243,043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02683797
CAS-Nummer: 51069-87-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR