3-(Carboxymethylaminomethyl)-4-hydroxybenzoic acid

APOSOR322055-1GEA 0 EUR
APOSOR322055-1G
3-(Carboxymethylaminomethyl)-4-hydroxybenzoic acid
3-(Carboxymethylaminomethyl)-4-hydroxybenzoic acid
Formel: C₁₀H₁₁NO₅
Molecular Weight: 225,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00134358
CAS-Nummer: 55739-39-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR