4-(Benzenesulphonyl)butan-2-one, tech

Lieferant: Apollo Scientific

APOSOR32201-1GEA 1390 EUR
APOSOR32201-1G
4-(Benzenesulphonyl)butan-2-one, tech
4-(Benzenesulphonyl)butan-2-one
Formel: C₁₀H₁₂O₃S
Molecular Weight: 212,265 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00243388
CAS-Nummer: 24731-39-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR