4-Chloro-N-methylbenzamide 95%

Lieferant: Apollo Scientific

APOSOR32198-1GEA 437 EUR
APOSOR32198-1G
4-Chloro-N-methylbenzamide 95%
4-Chloro-N-methylbenzamide
Formel: C₈H₈ClNO
Molecular Weight: 169,608 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00774485
CAS-Nummer: 6873-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR