N-[2-(1H-Pyrrol-1-yl)ethyl]acetamide 95%
Lieferant: Apollo Scientific
APOSOR32191-1GEA
1270
EUR
APOSOR32191-1G
N-[2-(1H-Pyrrol-1-yl)ethyl]acetamide 95%
N-[2-(1H-Pyrrol-1-yl)ethyl]acetamide
Formel:
C₈H₁₂N₂O Molecular Weight: 152,194 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00180783 CAS-Nummer: 73627-16-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...