(3E)-3-(Methoxyimino)-1-(4-methoxyphenyl)propan-1-one, tech
Lieferant: Apollo Scientific
APOSOR32177-1GEA
1390
EUR
APOSOR32177-1G
(3E)-3-(Methoxyimino)-1-(4-methoxyphenyl)propan-1-one, tech
(3E)-3-(Methoxyimino)-1-(4-methoxyphenyl)propan-1-one
Formel:
C₁₁H₁₃NO₃ Molecular Weight: 207,226 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00127145 CAS-Nummer: 477851-39-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...