3,4-Dihydro-2H-1,5-benzodioxepine-7-carboxylic acid 98%
|
Achtung
|
APOSOR23204-1GEA
0
EUR
APOSOR23204-1G
APOSOR23204-5G
3,4-Dihydro-2H-1,5-benzodioxepine-7-carboxylic acid 98%
3,4-Dihydro-2H-1,5-benzodioxepine-7-carboxylic acid
Formel:
C₁₀H₁₀O₄ Molecular Weight: 194,18 g/mol Schmelzpunkt: 145,2…147.3 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00276310 CAS-Nummer: 20825-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...