4-(Chlorocarbonyl)benzeneboronic anhydride
Lieferant: Apollo Scientific
|
Gefahr
|
APOSOR10418-1GEA
69.8
EUR
APOSOR10418-1G
APOSOR10418-5G
4-(Chlorocarbonyl)benzeneboronic anhydride
4-(Chlorocarbonyl)benzeneboronic anhydride
Formel:
C₇H₄BClO₂ Molecular Weight: 166,369 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06801674 CAS-Nummer: 380430-63-7 UN: 1759 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...