1-Phenyl-2,3-naphthalenedicarboxylic anhydride ≥98%, Sigma-Aldrich®
|
Achtung
|
456047-5GEA
307
EUR
456047-5G
1-Phenyl-2,3-naphthalenedicarboxylic anhydride ≥98%, Sigma-Aldrich®
1-Phenyl-2,3-naphthalenedicarboxylic anhydride
1-Phenyl-2,3-naphthalenedicarboxylic anhydride,98%
Formel:
C₁₈HO₃ Molecular Weight: 274,27 g/mol Schmelzpunkt: 259-262 °C (Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00030121 CAS-Nummer: 1985-37-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...