2-(Phenylsulfonyl)ethanol ≥97%, Sigma-Aldrich®

Lieferant: SIGMA-ALDRICH MICROSCOPY
Achtung

360686-5GEA 98.1 EUR
360686-5G
2-(Phenylsulfonyl)ethanol ≥97%, Sigma-Aldrich®
2-(Phenylsulfonyl)ethanol

2-(Phenylsulfonyl)ethanol,97%

Formel: C₈HO₃S
Molecular Weight: 186,23 g/mol
Dichte: 1,557 g/mL (25 °C) (Lit.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00134179
CAS-Nummer: 20611-21-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR