Disperse Orange 37, Sigma-Aldrich®

Lieferant: SIGMA-ALDRICH MICROSCOPY
Achtung

21603-5GEA 41 EUR
21603-5G
Disperse Orange 37, Sigma-Aldrich®
Disperse Orange 37

Disperse Orange 37 may be used during an experimental study conducted to understand the mutagenic and carcinogenic potential of textile azo dye processing plant effluent, that is known to have an impact on drinking water source.

Formel: C₁₇H₁₅Cl₂N₅O₂
Molecular Weight: 392,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00181820
CAS-Nummer: 13301-61-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR