Schisandrin B ≥96%

Lieferant: Thermo Scientific

ACRO465142500EA 340 EUR
ACRO465142500
Schisandrin B ≥96%
Schisandrin B

  • Bioactive

Formel: C₂₃H₂₈O₆
Molecular Weight: 400,46 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00210555
CAS-Nummer: 61281-37-6

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Powder
Infrared spectrum Conforms
Loss on drying ≤3 %
HPLC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR