FN1 motifs cell culture dishes, CCCadvanced®
Lieferant: Eppendorf
EPPE0038130.010EA 0 EUR
EPPE0038130.010 EPPE0038130.020
FN1 motifs cell culture dishes, CCCadvanced®
Ready-to-use and xeno-free: coated with synthetic RGD-derived motifs cell culture dishes for cultivation of stem cells (iPSCs, MSCs), primary cells or other ECM-dependent cells.
  • Pyrogen-, DNase-, RNase-, human and bacterial DNA-free
  • Non-cytotoxic
  • No tedious preparation with possible vessel/coating media dissipation
  • Suitable for many cell types, dissociation media (Accutase®, TrypLE™ reagents), as well as serum
  • Shelf life of 36 months from date of manufacture (stored dry at 15 to 37 °C)
  • No expensive lot-specific performance verification of coating media
  • Reduced contamination risk no preparation needed and several smart features for safer handling
  • High-quality base material, polystyrene (meets requirements of USP Class VI), manufactured with Quality Management System (QMS) compliant to standard QMS
  • Compatible with cell-based assays and staining procedures: no autofluorescence
CCCadvanced FN1 motifs consumables are additionally coated with a defined synthetic animal- and human-component-free substrate. The substrate (fibronectin-derived motifs with optimised steric configuration) is designed to mimic the cell attachment site of native ECM.
Applied for feeder-free cell culture and restrictive culture conditions (serum- and xeno-free). The SplashProtect™ ring inside the lid traps liquid and prevents spills and condensation. Gripping safe lid provided.