Sleeve protectors, BizTex® Microporous type PB[6], ST43
Lieferant: Portwest
BizTex®
PRTWST43WHREA 140 EUR
PRTWST43WHR
Sleeve protectors, BizTex® Microporous type PB[6], ST43
Einweg-Kleidung
The ST43 sleeve cover is breathable for wearer comfort. Elasticated at both ends to ensure no liquids or airborne particles can penetrate the barrier, this product has a much more relaxed fit.
  • Made of breathable microporous polypropylene/polyethylene
Standards: This fabric conforms to EN 14126 biological protection and EN 1149-1 anti-static protection, EN 13034 Type PB [6].
Bitte beachten Do not iron and wash.