Safety shoes, lace-up, Steelite™ WORK mid cut trainer, FW30
Lieferant: Portwest
Steelite™ WORK
PRTWFW30BKR47EA 70.9 EUR
PRTWFW30BKR47 PRTWFW30BKR39
Safety shoes, lace-up, Steelite™ WORK mid cut trainer, FW30
Schuhe und Stiefel
Classic trainer style with split leather upper and phylon/rubber sole resistant outsole to 300 °C.
  • Protective steel toecap
  • SRA - slip resistant outsole to prevent slips and trips on ceramic surfaces
  • Fuel and oil resistant outsole
  • Dual density sole unit
  • Slip, fuel and oil resistant rubber outsole
Colour: Black
Standards: EN ISO 20345, HRO