Rain Jackets, S534, Portwest
Lieferant: Portwest UC
PRTWS534BKRMEA 42.7 EUR
PRTWS534BKRM PRTWS534BKRXXL
Rain Jackets, S534, Portwest
Arbeitskleidung
An affordable safety jacket is ideal for anyone who is looking for a waterproof work jacket that is designed to last. Taped seams and two-parallel zipper cover hold air far away. Details such as cuffs, collar background and epolets make the jacket stand out, and classic.
  • Waterproof clothing protects in all weathers and keep warm
  • Fully lined and padded, fits to the cold
  • Taped seam for extra protection
  • Four spacious pockets
  • Phone pocket
  • Front slit
  • The double-acting chain is quick and convenient
  • Foldable hood for improved functionality
  • Internal elastic provide heat and protection
  • Quick-release radio loop radio
Shell fabric: 100% polyester, PU coated 130 g; lining fabric: 100% polyester 60 g; filling fabric: 100% polyester 120 g
Standards: EN 343:2019 Class 3:1 X (WP 5000 mm, MVP 1000 g/m²/24 h).