Face shield, CV15
Lieferant: Portwest
PRTWCV15CLREA 0 EUR
PRTWCV15CLR
Face shield, CV15
Gesichtsschutz
Full face shield designed to provide protection from fluid and splashes clear visor provides optimal visibility with clearance for prescription eyewear foam headband insert to improve comfort and fit for the user.
  • Padded headband for comfort
  • Anti-fog coating for additional comfort to the wearer
Portwest head PPE range applies the latest standards to deliver the highest level of protection. Comfortable and lightweight, the PPE range will ensure a pleasant wear, even for long periods of use.