Classic safety glasses, PW33
Lieferant: Portwest
Portwest® Classic
PRTWPW33AMREA 821 EUR
PRTWPW33AMR
Classic safety glasses, PW33
Schutzbrillen
Single panoramic lens, molded side shield and browguard for optimized protection against impacts, also suitable to wear over glasses.
  • 99% UV protection
  • Anti-scratch coating for added durability
  • Excellent side protection
  • Individually packed for vending machines
  • Adjustable length for a secure and comfortable fit
Standards: ANSI/ISEA Z87 + (U6 (amber)); EN 166
Lieferumfang: Cord included.