Wolfram(IV)carbid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Wolframcarbid

M-Clarity™ quality level = MQ100

SIAL241881-100GEA 246 EUR
SIAL241881-100G
Wolfram(IV)carbid, Sigma-Aldrich®
Wolfram(IV)carbid
Formel: CW
Molecular Weight: 195,85 g/mol
Siedepunkt: 6000 °C (1013 hPa)
Schmelzpunkt: 2785 °C
Dichte: 8,64 g/cm³ (20 °C)
MDL: MFCD00011464
CAS-Nummer: 12070-12-1
EINECS: 235-123-0
UN: 3178
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR