Tetraethylammoniumnitrat, Sigma-Aldrich®

Lieferant: Merck

SIAL86645-10GEA 73.2 EUR
SIAL86645-10G
Tetraethylammoniumnitrat, Sigma-Aldrich®
Tetraethylammonium nitrate
Formel: C₈H₂₀N₂O₃
Molecular Weight: 192,26 g/mol
Schmelzpunkt: > 200 °C
Dichte: 1,1622 g/cm³ (25 °C)
MDL: MFCD00041978
CAS-Nummer: 1941-26-0
EINECS: 217-725-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR