Tetrabutylammoniumcyanid, Sigma-Aldrich®

Lieferant: Merck
Gefahr

SIAL79989-100GEA 706 EUR
SIAL79989-100G
Tetrabutylammoniumcyanid, Sigma-Aldrich®
Tetrabutylammonium cyanide
Formel: C₁₇H₃₆N₂
Molecular Weight: 268,49 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 10442-39-4
EINECS: 233-926-0
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR