Tetrapropylammoniumtetrafluoroborat, Sigma-Aldrich®

Lieferant: Merck

SIAL88128-25GEA 180 EUR
SIAL88128-25G
Tetrapropylammoniumtetrafluoroborat, Sigma-Aldrich®
Tetrapropylammonium tetrafluoroborate
Formel: C₁₂H₂₈BF₄N
Molecular Weight: 273,16 g/mol
CAS-Nummer: 338-38-5
EINECS: 206-416-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR