Tetrahexylammoniumtetrafluoroborat, Sigma-Aldrich®

Lieferant: Merck

SIAL87315-10GEA 89.8 EUR
SIAL87315-10G
Tetrahexylammoniumtetrafluoroborat, Sigma-Aldrich®
Tetrahexylammonium tetrafluoroborate
Formel: C₂₄H₅₂BF₄N
Molecular Weight: 441,49 g/mol
CAS-Nummer: 15553-50-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR